Learn More
Description
- Dissociates cells while inactivating endogenous nucleases
- Lyses cultured cells directly
- Must be used in combination with a preliminary digest with Proteinase K for tissue samples
- Designed to be used with the Wizard SV or Wizard SV 96 Genomic DNA Purification System
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Isolation Technology | Alkaline Lysis |
| For Use With (Equipment) | Genomic DNA purification system |
| For Use With (Application) | Purifies genomic DNA from cultured cells and tissue |
| Product Type | Buffer |
| Quantity | 50 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.