Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ZAP70 Kinase Enzyme System

Catalog No. PRV8311 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
1000 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV8311 1000 Assays
PRV3811 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV8311 Supplier Promega Supplier No. V8311
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant full-length human ZAP70 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. ZAP70 is a nonreceptor protein tyrosine kinase (part of the Syk/Zap70 family) that is involved in signaling by the T-cell antigen receptor.

  • Easily Screen and Profile ZAP70 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Includes 1 x 1000 assays ADP-Glo™ Kinase assay and 1 x 10μg ZAP70 Kinase Enzyme system
Molecular Weight (g/mol) 96
Content And Storage -20°C to -70°C
Quantity 1000 Assays
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.