Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PRONASE™ Protease, Streptomyces griseus

Catalog No. 5370225KU Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10kU
250kU
25kU
500kU
50kU
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-702-25KU 25kU
53-702-10KU 10kU
53-702-50KU 50kU
53-702-250KU 250kU
53-702-500KU 500kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 53-702-25KU Supplier MilliporeSigma™ Supplier No. 5370225KU
Add to Cart
Add to Cart

Most active at neutral pH, but stable over wide range of pH and temperature

  • A non-specific protease liquefies mucins and digests proteins to free amino acids
  • Can be used to isolate living chondrocytes
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a solution containing calcium acetate as a stabilizer
Quantity 25kU
Format Lyophilized
pH Optimum 7.5
Content And Storage Store at+2°C to+8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.