Learn More
MilliporeSigma™ PRONASE™ Protease, Streptomyces griseus, Nuclease-Free
Shop All MilliporeSigma ProductsDescription
- Non-specific protease liquefies mucins and digests proteins to free amino acids
- Can be used to isolate living chondrocytes
- Most active at neutral pH, but stable over wide range of pH and temperature
Specifications
Specifications
| Quantity | 50kU |
| Format | Lyophilized |
| pH | Optimum 7.5 |
| Content And Storage | Store at+2°C to+8°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.