Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ProNex DNA QC Assay ABI 7500/7500FAST, 200 reactions

Catalog No. PRNG1002 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Content And Storage:
-30°C to -10°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
PRNG1002 -30°C to -10°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PRNG1002 Supplier Promega Supplier No. NG1002
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Evaluate quality and quantity of gDNA extracted from FFPE samples or other potentially degraded DNA sources with this human-specific, multiplexed probe-based qPCR assay. Also use to evaluate ratio of ccfDNA to higher molecular weight gDNA in plasma samples.

  • Evaluate Quality and Quantity of gDNA from Degraded Samples
  • Human−specific, multiplexed probe−based qPCR assay with internal positive control
  • Detect 75bp, 150bp and 300bp human genomic DNA sequences
  • Evaluate your samples for amplifiability and predict downstream assay success

Specifications

Content And Storage -30°C to -10°C
No. of Reactions 200
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.