Learn More
Promega ProNex DNA QC Assay Calibration Kit, 7500, 1 each
Catalog No. PRNG1001 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
ProNex DNA QC Assay Calibration Kit, 7500, 1 each
Catalog No.
PRNG1001 ProNex DNA QC Assay Calibration Kit, 7500, 1 each
1 options
This item is not returnable. View return policy
Catalog No. PRNG1001 Supplier Promega Supplier No. NG1001
This item is not returnable. View return policy
Evaluate quality and quantity of gDNA extracted from FFPE samples or other potentially degraded DNA sources with this human-specific, multiplexed probe-based qPCR assay. Also use to evaluate ratio of ccfDNA to higher molecular weight gDNA in plasma samples.
  • Evaluate Quality and Quantity of gDNA from Degraded Samples
  • Human−specific, multiplexed probe−based qPCR assay with internal positive control
  • Detect 75bp, 150bp and 300bp human genomic DNA sequences
  • Evaluate your samples for amplifiability and predict downstream assay success