Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ProNex Size-Selective Purification System, 500ml

Catalog No. PRNG2003 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRNG2003 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PRNG2003 Supplier Promega Supplier No. NG2003
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The ProNex Size-Selective Purification System enables the rapid and efficient magnetic resin-based purification of dsDNA for NGS, PCR and general molecular biology applications.

  • Superior Size Selection and Recovery for NGS and PCR
  • High specificity of size selection with low carryover of unwanted DNA
  • Exceptional DNA recovery
  • Fast magnetic response time and low viscosity for accurate pipetting

Specifications

Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.