Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propargylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. P099025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P099025G 25 g
P09905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P099025G Supplier TCI America Supplier No. P099025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15430-52-1
Molecular Formula C3H6ClN
Molecular Weight (g/mol) 91.54
MDL Number MFCD00012907
InChI Key IKXNIQJDNKPPCH-UHFFFAOYSA-N
Synonym propargylamine hydrochloride, prop-2-yn-1-amine hydrochloride, 2-propynylamine hydrochloride, propargylamine hcl, 2-propynylamine hcl, 3-amino-1-propyne hydrochloride, prop-2-ynylamine hydrochloride, 2-propyn-1-amine, hydrochloride, pubchem18845, acmc-1bvqh
PubChem CID 11205720
IUPAC Name hydrogen prop-2-yn-1-amine chloride
SMILES [H+].[Cl-].NCC#C

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 91.54
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Propargylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.