Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Properties of Materials Classroom Set
SDPSCI_ED

Catalog No. S27839 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Quantity:
Set of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S27839 Set of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27839 Supplier United Scientific Supplies Supplier No. PMTKITSET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allows students to make a wide range of investigations with only a small number of items

  • Kit allows students to make a wide range of investigations with only a small number of items
  • Contains two sets of three metal cylinders - aluminum, copper, and steel
  • One set of the cylinders have equal masses, and the other set of cylinders have equal volumes
  • The measuring instruments (cylinder, spring scale, and caliper) measure volume, mass, and length
  • Basic investigations include addressing the concepts of mass, volume, density, specific gravity, and extending to an exploration of buoyancy
  • Set of 5

Specifications

Quantity Set of 5
For Use With (Application) Measuring Properties of Materials
Product Type Properties of Materials Kit
Includes Two Sets of Three Metal Cylinders
Material Aluminum, Copper, Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.