Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Properties of Materials Kit
SDPSCI_ED

Catalog No. S00659 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S00659 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S00659 Supplier United Scientific™ Supplier No. PMTKIT
Add to Cart
Add to Cart

Properties of Materials Kit is a unique hands on kit for students.

This unique kit allows for a wide range of basic investigations, addressing the concepts of mass, volume, density, and specific gravity and extending to an exploration of buoyancy.

Contents:

  • 1 set of equal volume metal cylinders - aluminum, copper, steel
  • 1 set of equal mass metal cylinders - aluminum, copper, steel
  • 1 measuring cylinder, 100 mL
  • 1 spring scale, 2.5N/250 g
  • 1 vernier caliper, stainless steel

Specifications

Grade 6 to 12
Format Kit
Languages English
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.