Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionaldehyde diethyl acetal, 97%, Thermo Scientific™

Catalog No. AC175572500
Change view
Click to view available options
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC175572500 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC175572500 Supplier Thermo Scientific Chemicals Supplier No. 175572500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4744-08-5
Density 0.8280g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H16O2
Refractive Index 1.3884 to 1.3904 (20°C, 589nm)
Linear Formula C2H5CH(OC2H5)2
Quantity 250g
MDL Number MFCD00009245
Synonym hexyl methyl ether, hexane, 1-methoxy, methyl hexyl ether, ether, hexyl methyl, methyl-n-hexyl ether, n-hexyl methyl ether, unii-r99k6anq7t, methoxyhexane, r99k6anq7t, 2-oxaoctane
InChI Key MBNMGGKBGCIEGF-UHFFFAOYSA-N
SMILES CCOC(CC)OCC
IUPAC Name 1,1-diethoxypropane
Molecular Weight (g/mol) 132.20
PubChem CID 78484
Formula Weight 132.2
Percent Purity ≥96.0%
Chemical Name or Material Propionaldehyde diethyl acetal, 97%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-257-8
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.