Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionyl bromide, 95%

Catalog No. ac392360050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392360050 5 g Glass bottle
AC392360250 25 g Glass bottle
AC392361000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC392360050 Supplier Thermo Scientific Chemicals Supplier No. 392360050
Only null left
Add to Cart

CAS: 598-22-1 | C3H5BrO | 136.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-22-1
Molecular Formula C3H5BrO
Molecular Weight (g/mol) 136.98
MDL Number MFCD00000116
InChI Key RIBFXMJCUYXJDZ-UHFFFAOYSA-N
Synonym propionyl bromide, propanoyl-bromide, propionylbromide, propanyl bromide, acmc-209mg3, ksc270q9p, ribfxmjcuyxjdz-uhfffaoysa, inchi=1/c3h5bro/c1-2-3 4 5/h2h2,1h3
PubChem CID 69018
IUPAC Name propanoyl bromide
SMILES CCC(=O)Br

Specifications

Color Blue-Gray
Density 1.5200g/mL
Boiling Point 104°C
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4525 to 1.4575 (20°C, 589nm)
Linear Formula CH3CH2COBr
Quantity 5 g
Specific Gravity 1.52
Solubility Information Solubility in water: reacts violently
Formula Weight 136.98
Percent Purity 95%
Physical Form Tablets
Chemical Name or Material Propionyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-924-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.