Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ ProPolymer™ Flashlights

Catalog No. 19822943 Shop All Streamlight Products
Change view
Click to view available options
Battery Type:
2 AA Alkaline
4 AA Alkaline
C Alkaline
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Battery Type
19-822-943 4 AA Alkaline
19-819-920 C Alkaline
19-819-836 2 AA Alkaline
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-822-943 Supplier Streamlight™ Supplier No. 68251

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Waterproof, extremely tough, and incredibly bright

  • Polymer case with rubber face cap and unbreakable polycarbonate lens is waterproof and shock and corrosion resistant
  • One-handed push-button operation
  • Dual switch for constant-stream or momentary-blink operation
  • Xenon bulb

2AA ProPolymer:
Operates up to four hours on two AA alkaline batteries (included). Length: 6 in. (15.2cm). Weight: 3.7 oz. (105g). Color: Yellow.

3C ProPolymer:
Operates 5.5 hours on three C alkaline batteries (not included). Length: 8.5 in. (21.6cm). Weight: 13.5 oz. (383g). Color: Yellow.

4AA ProPolymer:
Operates up to four hours on four AA alkaline batteries (included). Length: 6.5 in. (16.5cm). Weight: 6.3 oz. (179g). Color: Yellow.

Order Info

Black fire helmet clamps available for models 2AA and 4AA.

Certifications

UL listed for certain Class I, Division 1 hazardous locations.

Specifications

Gas Fill Xenon
Weight (English) 6.3 oz.
Weight (Metric) 0.17 kg
Battery Type 4 AA Alkaline
Certifications/Compliance UL Class I, Division 1
Color Yellow
Controller Type On/Off
For Use With (Application) For constant-on or blink
Includes 4 AA batteries
Length (English) 6.5 in.
Length (Metric) 16.51 cm
Product Type Flashlight
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.