Learn More
Propper Mail-In Biological Indicator Test Pack and Culture Service
SDP
Catalog No. 0952541 Shop All Propper Manufacturing Co. Products
Change view
Click to view available options
Type:
Biological Test Pack
Catalog No. Type
09-525-41 Biological Test Pack
1 options
This item is not returnable. View return policy
Catalog No. 09-525-41 Supplier Propper Supplier No. 269067
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Biological indicators, pre-addressed postage paid envelopes and report forms.
Mail-In Biological Indicator Test Pack and Culture Service for daily or weekly monitoring of steam, ethylene oxide, or dry heat sterilizers. Easy-to-use mail-in service verifies and documents sterilizer performance (steam, dry heat, and EtO).
  • Mail-In Biological Indicator Test Pack and Culture Service Program makes biological monitoring easy for small medical, dental, and non-clinical offices.
  • A weekly or daily sterilizer monitoring and load monitoring during sterilization.
  • Mail-In Biological Indicator includes 10 pre-assembled biological indicator test packs for steam and ethylene oxide sterilization.
  • After processing, Test Pack is placed in a loaded sterilization chamber and processed, then mailed via prepaid return envelope.
  • In the event of a Failed test, a representative will reach out to you to inform you of your result.
Specifications
Type Biological Test Pack