Learn More
Propper Duo Spore™ Biological Indicator Test Envelopes
SDP
Catalog No. 0952540 Shop All Propper Manufacturing Co. Products
Change view
Click to view available options
Quantity:
25 Pk.
Catalog No. Quantity
09-525-40 25 Pk.
1 options
This item is not returnable. View return policy
Catalog No. 09-525-40 Supplier Propper Supplier No. 26906500
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Includes
Each set contains two test strips and one control strip.
Duo-Spore™ Biological Indicator Test for weekly or daily sterilizer monitoring and for load monitoring during sterilization. Self-contained biological indicator for use in steam or EO. Reliable results in just 48 hours. Suitable for sterilizers in all clinical and non-clinical settings.
  • Duo Spore™ Biological Indicator Strips monitor steam, ethylene oxide or dry heat sterilization cycles sterilization cycles in all clinical and non-clinical settings.
  • Prepared from Geobacillus stearothermophilus and Bacillus atrophaeus
  • Each test envelope contains 2 test strips and 1 control strip in two color separated pouches.
  • No Processing.
  • Process using Tryptic Soy Broth (TSB) culture media in your facility, which features a higher spore recovery rate, providing for a more dependable test.
  • The majority of all positive tests will occur within 24 hours for Steam and 48 hours for Ethylene Oxide.
  • Outgrowth rates may vary, depending on the source and type of media used.
  • The sub lethally damaged spores may require longer periods of incubation, up to 5 days.
  • D-values and Survival/Kill performance data supplied with each unit are in compliance with U.S. Pharmacopeia requirements.
Specifications
Product Type Biological Indicator
Includes 2 test strips and 1 control strip in each envelope
Quantity 25 Pk.
Sterility Non-sterile