Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propper Seracult™ Fecal Occult Blood Testing Triple Slides for Serial Testing
SDP

Catalog No. 1255427 Shop All Propper Manufacturing Co. Products
Change view
Click to view available options
Quantity:
34/Cs.
80/Cs.
Type:
Mailing Kit
Seracult Triple Slide
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
12-554-27 80/Cs. Mailing Kit
12-554-26A 34/Cs. Seracult Triple Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-554-27 Supplier Propper Supplier No. 37200700

missing translation for 'validMainframeMsg'

Seracult™ Guaiac Fecal Occult Blood Test is a qualitative aid in the diagnosis of colorectal cancer. Single or triple slide. Offers long sample stability and fast qualitative results.

  • Seracult™ Fecal Occult Blood Test is a qualitative aid in the diagnosis of colorectal cancer.
  • Designed for routine use in physical examinations in doctors' offices, hospitals, clinics, and laboratories or in the privacy and convenience of a patient's home.
  • Seracult™ Triple Slides made consecutive day testing easy and convenient for patients.
  • Seracult™ is available in a variety of configurations to meet all of your needs.
  • Results are ready instantly.
  • Seracult paper allows for enhanced readability for early detection of even the smallest amount of blood in the sample.
  • Detects hemoglobin and products of its degradation.
  • Mailing kits of in-office slides.
  • 28-day sample stability.
  • CLIA waived.

Specifications

Content And Storage Room Temperature
Format Kit, In vitro Diagnostic Test
Sample Type Stool
Type Mailing Kit
For Use With (Application) Fecal Occult Blood Detection
Includes Two Boxes, 40 Kits Ea. (Triple Slide and 3 Applicators, 3 Collection Tissues and USPS Approved Foil Lined Mailing Envelope) and 2 x 15 mL Bottles of Developer
Quantity 80/Cs.
Test Time 30 to 60 sec.

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.