Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propyl 3-Propoxypyridine-2-carboxylate 98.0+%, TCI America™
SDP

Catalog No. P11961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11961G 1 g
P11965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11961G Supplier TCI America Supplier No. P11961G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 134319-22-5
Molecular Formula C12H17NO3
Molecular Weight (g/mol) 223.272
MDL Number MFCD00191575
InChI Key ZGUCGLNLZXLQTD-UHFFFAOYSA-N
Synonym 3-Propoxypicolinic Acid Propyl Ester, Propyl 3-Propoxypicolinate, 3-Propoxypyridine-2-carboxylic Acid Propyl Ester
PubChem CID 19756120
IUPAC Name propyl 3-propoxypyridine-2-carboxylate
SMILES CCCOC1=C(N=CC=C1)C(=O)OCCC

Specifications

Color Yellow
Quantity 1 g
Formula Weight 223.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl 3-Propoxypyridine-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.