Learn More
Medchemexpress LLC Propyl 6-ethyl-5-ethylsulfanylcarbonyl-2-phenyl-4-propylpyridine-3-carboxylate | 212329-37-8 | >98.0% | 1 ML

Supplier: Medchemexpress LLC HY12111910MM 1ML
MRS 1523 is a selective adenosine A3 receptor antagonist used as a research reagent in pharmacology and neurophysiology studies. The manufacturer provides solvent and stock preparation guidance for analytical and biological use.
- Potent and selective adenosine A3 receptor antagonist; Ki ≈ 18.9 nM (human) and 113 nM (rat).
- Molecular formula C23H29NO3S; molecular weight 399.55 g/mol.
- CAS number 212329-37-8; commonly used as a research reagent and analytical standard.
- Datasheet includes solvent guidance and calculations for preparing 10 mM stock solutions.
- Purity reported as greater than 98% (HPLC) in depositor-supplied data.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.