Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propyl Alcohol n- 1-Propanol ACS, Greenfield

Catalog No. 502946017
Encompass
Change view
Click to view available options
Packaging:
Metal Drum
Metal Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
50-294-6017 Metal Drum
50-294-6060 Metal Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-294-6017 Supplier Thermo Scientific Production Chemicals and Services Supplier No. P334000ACSDM55M
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71-23-8
Molecular Weight (g/mol) 60.1

Specifications

Quantity 55 Gallon
Packaging Metal Drum
Boiling Point 206.6 °F (97 °C)
Percent Purity 100%
Grade ACS
Chemical Name or Material n-Propyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.