Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene Glycol (1,2-Propanediol), Reagents

Catalog No. 01337079
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-079 4 L
01-337-082 1 L
01-337-080 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-079 Supplier Reagents Holdings Llc Supplier No. C22933004C
Add to Cart
Add to Cart

Propylene Glycol (1,2-Propanediol), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 57-55-6
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.10
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
Synonym 1,2-Propanediol, Propane-1,2-diol
IUPAC Name propane-1,2-diol
SMILES CC(O)CO

Specifications

Boiling Range 188.2°C
Melting Point -60°C
Color Colorless
Boiling Point 188.2°C
Melting Range -60°C
Packaging Glass Bottle
Quantity 4 L
Specific Gravity 1.036
Viscosity Viscous
Grade Reagent
Physical Form Liquid
Chemical Name or Material Propylene Glycol (1,2-Propanediol)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.