Learn More
Sigma Organic Chemistry (+/-)-Propylene oxide | 250ML | 75-56-9 | MFCD00005126 | >=99.5% (GC)
Supplier: Sigma Organic Chemistry 82320250ML

(+/-)-Propylene oxide, 250ML
About This Item:
Synonym(s): (+/-)-Methyloxirane; 1,2-Epoxypropane
Linear Formula: C3H6O
Molecular Weight: 58.08
CAS Number: 75-56-9
MDL Number: MFCD00005126
UNSPSC Code: 12162002
Purity: >=99.5% (GC)
Boiling Point: 34 C (lit.)
Melting Point: -112 C (lit.)
Density: 0.83 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.366 (lit.); n20/D 1.366
Storage: room temp
EINECS Number: 200-879-2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.