Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Propylene oxide, >99%

Catalog No. p-7049891
Change view
Click to view available options
Quantity:
100 mL
1 L
3 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA30765K2 1 L
AA30765L7 3 x 1 L
AA30765AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA30765K2 Supplier Thermo Scientific Chemicals Supplier No. 030765K2
Only null left
Add to Cart

CAS: 75-56-9 | C3H6O | 58.08 g/mol

Propylene oxide is used as a precursor in the production of polyether polyols, which is utilized for making polyurethane plastics, polypropylene glycols, propylene glycol ethers and propylene carbonate. It is involved in the preparation of biological samples for electron microscopy. It plays a vital role in the removal of residual ethanol previously used for dehydration. Further, it is used in thermobaric weapons, and microbial fumigation. It is also used as solvents, and in the preparation of surfactants, lubricants and in oil demulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-56-9
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00005126
InChI Key GOOHAUXETOMSMM-UHFFFAOYNA-N
Synonym propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
PubChem CID 6378
ChEBI CHEBI:38685
IUPAC Name 2-methyloxirane
SMILES CC1CO1

Specifications

Melting Point -112°C
Color Colorless
Density 0.83 g/mL
Boiling Point 34°C to 35°C
Flash Point -37°C (-35°F)
Odor Ether-like
Assay Percent Range >99%
Refractive Index 1.366
Quantity 1 L
UN Number UN1280
Beilstein 79763
Merck Index 14,7856
Solubility Information Miscible with water,acetone,benzene,carbon tetrachloride,methanol and ether.
Formula Weight 58.08
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material (+/-)-Propylene oxide
Show More Show Less
Hazard Category H224-H302-H311+H331-H319-H335-H340-H350
Hazard Statement GHS H Statement
H224-H340-H350-H302-H312-H332-H315-H319-H335
Extremely flammable liquid and vapor.
May cause genetic defects.
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: PROPYLENE OXIDE
EINECSNumber 200-879-2
RTECSNumber TZ2975000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.