Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Propylene Oxide, Analytical Standard, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019167 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9167 5 mL
11-102-0591 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9167 Supplier MilliporeSigma Supelco Supplier No. 566715MLF
Add to Cart
Add to Cart

Specifications

CAS 75-56-9
Vapor Pressure 8.59 psi (20°C)
Vapor Density 2 (vs. air)
Density 0.83 g/mL (at 25°C)
Physical Form Neat
Linear Formula C3H6O
MDL Number MFCD00005126
Melting Point -112°C
Boiling Point 34°C
Synonym (+/-)-Methyloxirane; 1,2-Epoxypropane
Quantity 5 mL
Formula Weight 58.08
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.