Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene Sulfide (stabilized with Butyl Mercaptan) 98.0+%, TCI America™
SDP

Catalog No. P032125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P032125ML 25 mL
P03215ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P032125ML Supplier TCI America Supplier No. P032125ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1072-43-1
Molecular Formula C3H6S
Molecular Weight (g/mol) 74.14
MDL Number MFCD00005160
InChI Key MBNVSWHUJDDZRH-UHFFFAOYNA-N
Synonym propylene sulfide, thiirane, methyl, thiirane, 2-methyl, propene sulfide, 1,2-epithiopropane, propylene sulphide, propylene episulfide, 2-methylthiacyclopropane, propane, 1,2-epithio, methylthiirane
PubChem CID 14072
IUPAC Name 2-methylthiirane
SMILES CC1CS1

Specifications

Color Yellow
Boiling Point 75°C
Quantity 25 mL
UN Number 1992
Formula Weight 74.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propylene Sulfide (stabilized with Butyl Mercaptan)
RTECSNumber TZ2625000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.