Learn More
Description
Propylparaben NF is a small, colorless crystals or white powder.
Alternative name(s): propyl hydroxybenzoate, Propyl paraben, Propyl parahydroxybenzoate
Specifications
Specifications
| Chemical Name or Material | Propylparaben NF |
| Packaging | Jar, 300mL, White, HDPE, Round, 63-400, TE Pano Neck |
| CAS | 94-13-3 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 180.202 |
| Quantity | 100 g |
| Percent Purity | 98 to 102 % |
| Grade | NF |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
