Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylthiouracil, USP, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18605939 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-939 5 g Amber Glass Bottle
18-605-943 5 kg Poly Bottle
18-605-940 25 g Amber Glass Bottle
18-605-941 100 g Amber Glass Bottle
18-605-942 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-605-939 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P20755GM

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

P2075, 51-52-5, C7H10N2OS

Spectrum™ Chemical Propylthiouracil, USP is an anti-thyroid drug used to manage hyperthyroidism which is due to an overactive thyroid gland. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 51-52-5
Molecular Formula C7H10N2OS
Molecular Weight (g/mol) 170.23
MDL Number MFCD00006041
InChI Key KNAHARQHSZJURB-UHFFFAOYSA-N
IUPAC Name 6-propyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
SMILES CCCC1=CC(=O)NC(=S)N1

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Melting Range 218-221°C
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 98.0 to 100.5%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.