Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-1 Single Domain Antibody [F42D7], ProSci
SDP

Catalog No. 16595651 Shop All ProSci Products
Change view
Click to view available options
Quantity:
0.1 mg
20 μg Trial Size
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-651 20 μg Trial Size
16-595-328 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-595-651 Supplier ProSci LLC Supplier No. SD86450020
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Llama Recombinant Monoclonal Antibody

Specifications

Antigen PD-1 Single Domain
Applications ELISA, Flow Cytometry
Classification Recombinant Monoclonal
Clone F4-2D7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution PD-1 antibody can be used for detection of PD-1 by ELISA and FACS (10 μg/ml).
Formulation PD-1 Antibody is supplied in PBS.
Gene Accession No. NP_005009
Gene Alias PD-1 Antibody: PD1, PD-1, CD279, SLEB2, hPD-1, hPD-l, hSLE1, PD1, Programmed cell death protein 1, Protein PD-1, PDCD1, PDCD-1
Host Species Llama
Purification Method PD-1 Antibody is affinity chromatography purified via Nickel column. Antibody is supplied as a His-tagged purified protein. It also contains a myc-tag for detection.
Quantity 20 μg Trial Size
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5133
Target Species Human
Content And Storage PD-1 antibody should be stored in working aliquots at -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Product Type Antibody
Form Liquid
Isotype Not Applicable
Research Category Immunology,Cancer,Cell Cycle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.