Learn More
AdipoGen Proquinazid
SDP
Supplier:  AdipoGen CDXP0126G001
Encompass_Preferred
Chemical. CAS 189278-12-4. Formula C14H17IN2O2. MW 372.2. Synthetic Choline fungicide that inhibits spore germination of fungal spores by preventing the formation of appressoria that are necessary for infection of host plants. Possible secondary mode of action is stimulating the expression of host defence genes. Controls powdery mildew on cereals and grapevines. Compound can be used as analytical reference material.
Catalog No. 50-256-0239
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.