Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PROS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7207453 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1252 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1252 Supplier Proteintech Group Inc Supplier No. 169101AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a vitamin K-dependent plasma protein that functions as a cofactor for the anticoagulant protease, activated protein C (APC) to inhibit blood coagulation. It is found in plasma in both a free, functionally active form and also in an inactive form complexed with C4b-binding protein. Mutations in this gene result in autosomal domit hereditary thrombophilia. An inactive pseudogene of this locus is located at an adjacent region on chromosome 3.

Specifications

Antigen PROS1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PROS1
Gene Accession No. P07225, Q08761
Gene Alias PROS, PROS1, protein S (alpha), PS21, PS22, PS23, PS24, PS25, PSA, Vitamin K dependent protein S
Gene Symbols Pros1
Host Species Rabbit
Immunogen PROS1 Fusion Protein Ag10539
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19128, 5627
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.