Learn More
CPC Scientific H-Thr-D-Ala-Leu-Ile-Asp-Asn-Asn-Ala-Thr-Glu-Glu-Ile-Leu-Tyr-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific PROS001B
Encompass
This 14-mer prosaptide sequence is derived from the active neurotrophic region in the amino-terminal portion of the saposin C domain. Synthetic peptides derived from this region are biologically active and are named ?prosaptides.? Prosaposin and prosaptides are active on a variety of neuronal cells, stimulating sulfatide synthesis and increasing sulfatide concentration in Schwann cells and oligodendrocytes. This indicates that prosaposin and prosaptides are trophic factors for myelin formation.ONE-LETTER SEQUENCE: TaLIDNNATEEILYMOLECULAR FORMULA: C69H110N16O26MOLECULAR WEIGHT:1579.70STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [196391-82-9], RESEARCH AREA: NeuropeptidesREFERENCES: Campana, M. et al. FASEB J. 12, 307 (1998); Hiraiwa, M. et al. Proc. Natl. Acad. Sci. USA 94, 4778 (1997); Taylor, E. et al. Pharmacol. 295, 190 (2000); Calcutt, NA. et al. Anesthesiology 93, 1271 (2000).
Catalog No. 50-211-0330
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.