Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Prosaptide TX14(A)
GREENER_CHOICE

Catalog No. 51511
Change view
Click to view available options
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
51-511 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 51-511 Supplier Tocris Bioscience™ Supplier No. 5151/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent GPR37 and GPR37L1 agonist

Prosaptide TX14(A) is a potent GPR37L1 and GPR37 agonist (EC50 values are 5 and 7 nM, respectively). Stimulates signaling through pertussis toxin-sensitive G proteins. Stimulates ERK phosphorylation. Protects against cellular stress. Promotes myelin lipid synthesis and prolongs cell survival in both Schwann cells and oligodendrocytes; neuroprotective and glioprotective.

Specifications

Purity 95%
Molecular Weight (g/mol) 1579.72
Product Type Prosaptide TX14(A)
Quantity 1 mg

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.