Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes
BIOPHARMAGREENER_CHOICE

Catalog No. p-2436379 Shop All Contec Inc. Products
Change view
Click to view available options
Dimensions (L x W):
22.86 x 22.86 cm (9 x 9 in.)
27.9 x 23 cm (11 x 9 in.)
Packaging Quantity:
24 Pk.
40 Packs of 20 Wipes
50 Pk.
Saturation Solution:
70% IPA, 30% DI Water
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Packaging Quantity Saturation Solution
19-150-0710 27.9 x 23 cm (11 x 9 in.) 24 Pk. 70% IPA, 30% DI Water
19-150-0706 27.9 x 23 cm (11 x 9 in.) 50 Pk. 70% IPA, 30% DI Water
19-170-356 22.86 x 22.86 cm (9 x 9 in.) 40 Packs of 20 Wipes 70% IPA, 30% DI Water
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-150-0710 Supplier Contec™ Supplier No. PSC20001
Add to Cart
Add to Cart
This item is not returnable. View return policy

Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for sterile environments.

  • Designed for pharmaceutical, biotech, and medical device applications
  • Choose spunlaced nonwoven 100% polyester wipes for improved cleanliness, economical hydroentangled polyester/cellulose wipers, or meltblown polypropylene wipes for improved durability
  • Conveniently presaturated with 70/30 blend of semi-conductor grade IPA and DI water to reduce VOC emissions and hazardous waste
  • All wipes are sterilized by gamma radiation to a 10-6 Sterility Assurance Level and validated per AAMI ISO 11137 (Method 1) guidelines
  • Certificate of Sterility and Certificate of Irradiation included
  • Environment: ISO 5-8, Grade A/B
  • Individually lot numbered resealable pouches with tamper-evident labels ensure product integrity and cleanliness

Compliance

Validated sterile to a Sterility Assurance Level of 10-6 in accordance with AAMI/ISO 1137 Method 1 Guidelines.

Specifications

Product Type Pre-saturated Wipe
Length (English) 9 in.
Length (Metric) 23 cm
Width (English) 11 in.
Width (Metric) 27.9 cm
Material Polyester Cellulose
Certifications/Compliance ISO Class 5-8
Dimensions (L x W) 27.9 x 23 cm (11 x 9 in.)
For Use With (Application) Ideal for routine cleaning and wipe down of lab tools, instruments and other equipment and designed for sterile environments
Packaging Quantity 24 Pk.
Saturation Solution 70% IPA, 30% DI Water
Sterility Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.