Learn More
PROSC Mouse anti-Human, Clone: OTI3A6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505163

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Pyridoxal 5′-phosphate (PLP)-binding protein, which may be involved in intracellular homeostatic regulation of pyridoxal 5′-phosphate (PLP), the active form of vitamin B6.Specifications
PROSC | |
Monoclonal | |
Unconjugated | |
PLPBP | |
EPVB6D, PROSC | |
Mouse | |
Affinity Chromatography | |
RUO | |
11212 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI3A6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O94903 | |
PLPBP | |
Full length human recombit protein of human PROSC produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.