Learn More
Electron Microscopy Sciences ProScope ProScope T1 WiFi Microscope 50x, 200x lens
SDP
Catalog No. 501931654
Encompass
Powered by a rechargable battery, ProScope T1 WiFi Microscope creates its own Wi-Fi network. It can transmit images simultaneously to as many as 253 devices (IOS or Android). With these apps, users can view live images, freeze and capture them to the photo album in their device. Ideal for dermatology, trichology and dental use.
Catalog No. 50-193-1654 Supplier Electron Microscopy Sciences Supplier No. 68350T1
Powered by a rechargable battery, ProScope T1 WiFi Microscope creates its own Wi-Fi network. It can transmit images simultaneously to as many as 253 devices (IOS or Android). With these apps, users can view live images, freeze and capture them to the photo album in their device. Ideal for dermatology, trichology and dental use.