Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 50 Coveralls with Hood

Catalog No. 19065503C Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Size (English) Chest:
29.75 to 33.25 in.
33.25 to 37.25 in.
37.25 to 40.75 in.
40.75 to 44.25
44.25 to 47.75 in.
47.75 to 51.25 in.
51.25 to 54.75 in.
54.75 to 58.25 in.
57.25 to 60.75 in.
Height (English):
5 to 5.58 ft.
5.22 to 6.16 ft.
5.25 to 5.58 ft.
5.41 to 5.75 ft.
6 to 6.33 ft.
6.16 to 6.33 ft.
6.33 to 6.58 ft.
6.58 to 6.83 ft.
6.75 to 7.08 ft.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Size (English) Chest Height (English)
19-065-503C Large 37.25 to 40.75 in. 5.41 to 5.75 ft.
19-065-503A Small 29.75 to 33.25 in. 5 to 5.58 ft.
19-065-503B Medium 33.25 to 37.25 in. 5.25 to 5.58 ft.
19-065-503D X-Large 40.75 to 44.25 5.22 to 6.16 ft.
19-065-503E 2X-Large 44.25 to 47.75 in. 6 to 6.33 ft.
19-065-503F 3X-Large 47.75 to 51.25 in. 6.16 to 6.33 ft.
19-065-503G 4X-Large 51.25 to 54.75 in. 6.33 to 6.58 ft.
19-065-503H 5X-Large 54.75 to 58.25 in. 6.58 to 6.83 ft.
19-065-503I 6X-Large 57.25 to 60.75 in. 6.75 to 7.08 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Should not be used around heat, flame, sparks or potentially flammable or explosive environments.
Catalog No. 19-065-503C Supplier DuPont™ Supplier No. NB127SWHLG002500
Add to Cart
Add to Cart
This item is not returnable. View return policy
Should not be used around heat, flame, sparks or potentially flammable or explosive environments.

Microporous film coveralls provide protection from non-hazardous aerosols, light liquid splash, and dry particles.

Microporous film coveralls provide protection from non-hazardous aerosols, light liquid splash, and dry particles.

  • Made with a microporous film that is laminated to a spundbonded polypropylene substrate
  • Front zipper closure
  • Elastic wrists, ankles
  • Attached hood
  • Serged seams
  • Collar
  • White
  • Thickness: 10 mils
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material ProShield
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Standard, Elastic Face
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Model NB127S
Size (English) Chest 37.25 to 40.75 in.
Size (English) Inseam 27.5 in.
Size (Metric) Inseam 69.85 cm
Thickness 10 mil
For Use With (Application) Janitorial, Sanitation, General Industrial, Maintenance
Length (English) Sleeve 33.25 in.
Length (Metric) Sleeve 84.46 cm
Width (English) Chest 25.25 in.
Width (Metric) Chest 64.135 cm
Height (English) 5.41 to 5.75 ft.
Height (Metric) 1.65 to 1.75 m
Quantity 25/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.