Learn More
DuPont™ PROSHIELD™ 6SFR Coverall with Standard Fit Hood
Shop All DuPont Personal Protection ProductsDescription
DuPont™ ProShield® 6 SFR garments are cost-efficient and designed to be worn over primary flame resistant clothing.
- Keeps dirt and grime off of primary flame resistant garments
- Provides a barrier against non-hazardous particles and aerosols
Specifications
Specifications
| Size | Medium |
| Gender | Unisex |
| Color | Blue |
| Disposable | Single-use |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Hood Type | Standard |
| Seam Type | Serged |
| Sterility | Non-sterile |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.