Learn More
Description
DuPont ProShield 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Garments are available in a variety of coverall styles. Applications include: general maintenance, janitorial/cleaning and other dirty work.
- Frock provides body-length jacket design with front zipper closure
- Mandarin collar
- Extends below hip
- Elastic wrists
- Serged seams
- Manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
| Size | 4X-Large |
| Material | ProShield |
| Color | White |
| Disposable | Single-use |
| Closure Type | Zipper |
| Cuff Style | Elastic |
| Length (Metric) | 104.1 cm |
| Length (English) | 41 in. |
| Product Type | General Purpose Frock |
| Seam Type | Serged |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.