Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProsTaglandin E Receptor EP4 Polyclonal Antibody, FITC, Bioss
SDP

Catalog No. 502280370
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-0370 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-0370 Supplier Bioss Supplier No. BS24918RFITC
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ProsTaglandin E Receptor EP4 Polyclonal antibody specifically detects ProsTaglandin E Receptor EP4 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen ProsTaglandin E Receptor EP4
Applications Flow Cytometry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate FITC
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene PTGER4
Gene Accession No. P32240
Gene Alias EP1; EP4; EP4R; MGC126583; PGE receptor EP4 subtype; PGE receptor, EP4 subtype; PGE2 receptor EP4 subtype; prostaglandin E receptor 4; prostaglandin E receptor 4 (subtype EP4); prostaglandin E2 receptor EP4 subtype; prostaglandin E2 receptor type 4; prostanoid EP4 receptor; Ptger; PTGER2; Ptger4; Ptgerep4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse EP4
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19219
Target Species Mouse
Content And Storage -20° C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.