Learn More
Enzo Life Sciences Prostaglandin E1, (10mg), CAS Number: 745-65-3
SDP
Supplier:  Enzo Life Sciences BMLPG0060010
Encompass_Preferred
Prostanoid receptor ligand. Biosynthesis from dihomo-γ-linolenic acid. A major prostaglandin in semen. Inhibits human platelet aggregation. Effects induced by receptor-mediated elevation of cAMP. Alternative Name: PGE1. Formula: C20H34O5. MW: 354.5. CAS Number: 745-65-3. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in ethanol (50mg/ml) or acetone (10mg/ml). Long Term Storage: -20°C
Catalog No. NC0829224
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.