Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prostaglandin I2 Sodium Salt, MP Biomedicals™

Catalog No. MP215196905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215196905 5 mg
MP215196910 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP215196905 Supplier MP Biomedicals, Inc Supplier No. 0215196905
Only null left
Add to Cart
Add to Cart

Inhibits platelet aggregation by antagonizing thromboxane A. Prostaglandin I2 Sodium Salt, MP Biomedicals is a short-lived product of the cyclooxygenase pathway in vascular endothelial cells. Prostacyclin therapy improves hemodynamics in pulmonary arteria

Prostaglandin I2 Sodium Salt is a potent fibrinolytic that antagonizes thromboxane A. It relaxes vascular smooth muscle and induces vasodilation. It also increases cyclic AMP concentration in human platelets (IC50 for human platelet aggregation is about 0.001 nM). Prostacyclin is vasoprotective, protecting arterial walls from injury-induced lesions and cytoprotective in the liver and gastrointestinal tract.

Prostaglandin I2 sodium salt stimulates platelet adenylyl cyclase. Nitric oxide is also produced in vascular endothelium, where it inhibits platelet aggregation, regulates inducible cyclooxygenase production, and may work synergistically with prostacyclin to attenuate the thrombotic process.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61849-14-7
Molecular Formula C20H31NaO5
Molecular Weight (g/mol) 374.453
InChI Key LMHIPJMTZHDKEW-HIHHSTLZSA-M
PubChem CID 133687047
IUPAC Name sodium;(5Z)-5-[(3aS,4S,5S,6aR)-5-hydroxy-4-[(E,3R)-3-hydroxyoct-1-enyl]-3,3a,4,5,6,6a-hexahydrocyclopenta[b]furan-2-ylidene]pentanoate
SMILES CCCCCC(C=CC1C(CC2C1CC(=CCCCC(=O)[O-])O2)O)O.[Na+]

Specifications

Quantity 5 mg
Solubility Information PGI2 is a hygroscopic crystalline solid soluble in water.
Formula Weight 374.5
Percent Purity ≥99%
Physical Form Powder
Recommended Storage −20°C. Store Desiccated.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.