accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaCH3(S)HO(R)(R)HO(S)O(R)OO(E)(Z)

CAS RN 61849-14-7

IUPAC Name: sodium 5-[(2Z,3aR,4R,5R,6aS)-5-hydroxy-4-[(1E,3S)-3-hydroxyoct-1-en-1-yl]-hexahydro-2H-cyclopenta[b]furan-2-ylidene]pentanoate
Synonyms: sodium prostaglandin I2(1-)
Molecular Weight (g/mol): 374.45
Molecular Formula: C20H31NaO5
InChi Key: LMHIPJMTZHDKEW-XQYLJSSYSA-M
SMILES: [Na+].CCCCC[C@H](O)\C=C\[C@H]1[C@H](O)C[C@@H]2O\C(C[C@H]12)=C/CCCC([O-])=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results