Learn More
Apexbio Technology LLC Immunoglobulin M heavy chain (IGHM) fragment [Homo sapiens] 5mg
SDP
Supplier:  Apexbio Technology LLC A10735
Encompass_Preferred
Immunoglobulin M heavy chain IGHM fragment is a peptide derived from the constant (C-terminal) region of the human immunoglobulin mu heavy chain It is designed for use in studies investigating immunoglobulin structure-function relationships and understanding antigen-binding mechanisms in the context of antibody-mediated immune responses Immunoglobulin M heavy chain IGHM fragment exerts its biological utility by serving as a research tool to elucidate the roles of the IgM constant region in antigen recognition and primary immune response initiation Based on these properties Immunoglobulin M heavy chain IGHM fragment holds research potential in biomedical fields such as immunoglobulin structure-function studies analysis of antigen-binding mechanisms and the development of immunodiagnostic assays
Catalog No. 50-000-96895
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.