Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Therapak™ Prostate Biopsy Collection and Transport Kit

Catalog No. 22131459 Shop All Therapak Corporation Products
Change view
Click to view available options
Holds:
12 vials
16 vials
8 vials
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
22-131-459 16 vials
22-130-056 12 vials
22-131-457 8 vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-131-459 Supplier Therapak™ Supplier No. 38871
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Complete with your choice of 6, 8, 12, 14 or 16 prefilled jars with 10mL formalin solution

Specifications

Product Type Biopsy Collection and Transport Kit
Sterility Non-sterile
Capacity (Metric) 20 mL
Closure Type Screw Cap
Graduations None
Holds 16 vials
Includes 10mL formalin
No. per Case 20/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.