Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prostate Specific Acid Phosphatase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143871 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143871 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143871 Supplier Invitrogen™ Supplier No. PA5143871
  • $447.00
    $415.65 / Each of 1

    Save $31.35

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human MCF-7 whole cell. IHC: human prostate cancer tissue, mouse kidney tissue, rat kidney tissue. ICC/IF: U87 cell. Flow: HL-60 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

ACPP is an enzyme which catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is synthesized under androgen regulation and is secreted by the epithelial cells of the prostate gland.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prostate Specific Acid Phosphatase
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene ACP3
Gene Accession No. P15309, P20646, Q8CE08
Gene Alias 5'-NT; 5'-nucleotidase; A030005E02Rik; acid phosphatase, prostate; ACP3; ACP-3; Acpp; Acpp11; EC 3.1.3.2; Ecto-5'-nucleotidase; fluoride-resistant acid phosphatase; FRAP; Lap; lysosomal acid phosphatase; PAP; PAPf39; Ppal; PPAP; prostate acid phosphatase; Prostatic acid phosphatase; prostatic acid phosphatase (rPAP); prostatic acid phosphotase; RNACPP11; Thiamine monophosphatase; TMPase
Gene Symbols ACP3
Host Species Rabbit
Immunogen E.coli-derived human PAP/ACP3 recombinant protein (Position: E150-D386).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55, 56318, 56780
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.