Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Prostratin, PKC Activator, Solid, 1 mg

Catalog No. 502707177
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7177 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7177 Supplier Abcam Supplier No. AB1208801MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent PKC activator that reactivates HIV-1 latency via NF-κB activation and downregulates CD4, CXCR4, and CCR5 expression.

  • Molecular Weight: 390.48 g/mol
  • Biological Activity: PKC activator; reactivates HIV-1 latency (IC50 = ~0.5 μM) via PKC-dependent NF-κB activation
  • Mechanism: Downregulates expression of HIV-1 receptor CD4 and its co-receptors CXCR4 and CCR5 in vitro
  • Characteristics: Non-tumor promoting
  • Form: Solid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at -20°C.
Form Solid
Target Molecule Protein Kinase C
Shipping Condition Blue Ice
Formulation 100% Prostratin
Quantity 1 mg
For Use With (Application) PKC activation, HIV-1 latency reactivation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.