Learn More
Medchemexpress LLC HY-111848A 5mg Medchemexpress, PROTAC AR Degrader-4 (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY111848A5MG
Encompass_Preferred
Medchemexpress, HY-111848A 5mg PROTAC AR Degrader-4 (TFA) CAS: PROTAC AR Degrader-4 comprises a cIAP1 ligand binding group, a linker and an androgen receptor (AR) binding group. PROTAC AR Degrader-4 is an AR degrader. Degradation inducers based on cIAP1 are called specific and non-genetic IAP-dependent protein erasers (SNIPERs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0804
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.