Learn More
Medchemexpress LLC PROTAC BCR-ABL Degrader-1 | Molecular Weight 792.84 | Formula C43H40N10O6 | Purity: 98.09% | 10 mg
SDP
Supplier:  Medchemexpress LLC HY15341510MG
Encompass_Preferred
PROTAC BCR-ABL Degrader-1 (compound PROTAC 1) is a PROTAC with a 2-oxoethyl linker. PROTAC BCR-ABL Degrader-1 induces Bcr-Abl degradation in a ubiquitinproteasom-dependent manner. PROTAC BCR-ABL Degrader-1 exhibits antiproliferative activity against K562 cells, and has the potential to study cancer.
Catalog No. NC2832497