Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,E)-N-(4-(2-((6-(2-(4-(4-Chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f]triazolo[4,3-a]diazepin-6-yl)acetamido)hexyl)amino)-2-oxoethoxy)benzyl)-2-cyano-3-(naphthalen-2-yl)acrylamide | 98.1% | 867.46 | C48H47ClN8O4S | 10 MG
SDP

Supplier:  Medchemexpress LLC HY15876410MG

Encompass_Preferred

PROTAC BET Degrader-12 is a proteolysis targeting chimera (PROTAC) designed to selectively degrade bromodomain and extra-terminal (BET) family proteins such as BRD3 and BRD4. Supplied as a research reagent in small-mass packs, it is intended for use in targeted protein degradation and epigenetics studies.

  • Selective BET protein degrader targeting BRD3 and BRD4.
  • Purity 98.1% by HPLC.
  • Molecular formula C48H47ClN8O4S; molecular weight 867.46.
  • Available in small-mass packs (e.g., 10 mg) for research use.
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for human or diagnostic use.

Catalog No. 50-002-88277


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.