Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PROTAC BET degrader- 1mg | 3031993-61-7 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY1142291MG

Encompass_Preferred

PROTAC BET degrader-3 is a proteolysis-targeting chimera that links a von Hippel-Lindau (VHL) ligand to a BET ligand to induce targeted proteasomal degradation of BET proteins. It is supplied as a light-yellow solid for research use.

  • Induces VHL-mediated degradation of BET proteins.
  • Molecular formula C53H64N12O9S; molecular weight 1045.22 g·mol⁻¹.
  • Reported purity 99.11%.
  • Available in small milligram quantities for screening and cell-based studies.
  • Store powder at -20°C; in solution store at -80°C for long term.

Catalog No. 50-002-29804


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.