Learn More
Medchemexpress LLC PROTAC BET degrader-1 | 2093386-22-0 | 99.4% | 871.90 g·mol⁻¹ | C44H45N11O9 | 25 MG
Supplier: Medchemexpress LLC HY10363325MG
PROTAC BET Degrader-1 is a proteolysis-targeting chimera that links Cereblon and BET ligands to induce degradation of BET family proteins BRD2, BRD3, and BRD4 at low concentrations. It is supplied as a light yellow solid for research use only.
- Degrades BRD2, BRD3, and BRD4 at low nanomolar concentrations.
- High purity suitable for biochemical and cell-based assays.
- Readily soluble in DMSO; protocols provided for in vivo formulations.
- Stable when stored as powder at -20 °C and in solvent at -80 °C.